missing translation for 'onlineSavingsMsg'
Learn More

Polyethylene Glycol 15-Hydroxystearate (Technical Grade, contains up to 35% polyethylene glycol), TRC

Product Code. 30427136
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427136 50 g 50g
30411397 100 g -
30410555 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427136 Supplier Toronto Research Chemicals Supplier No. TRCP68852050G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70142-34-6
Molecular Formula C18H36O3(C2H4O)n
Molecular Weight (g/mol) 362.5445
Synonym Macrogol (15)-Hydroxystearate, Kolliphor HS15, Solutol HS15
SMILES OC(CCCCCCCCCCC(O)CCCCCC)=O.[H]OCCO

Specifications

Quantity 50 g
Formula Weight 362.3032
Chemical Name or Material Polyoxyl 15 Hydroxystearate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.