Learn More
Polyethylene glycol 12,000
Description
Polyethylene glycol is used to modify therapeutic proteins and peptides to increase their solubility. In gas chromatography, it is used as a polar stationary phase. It is also useful in preserving objects obtained from the ocean. As a dispersant, it is used in toothpastes. It finds application in blood banking as a potentiator, which is used to detect antigens and antibodies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| pH | 5 to 7 |
| Physical Form | Powder |
| Melting Point | 64°C to 65°C |
| Merck Index | 14,7568 |
| Solubility Information | Soluble in organic solvents,water and aromatic hydrocarbons. Slightly soluble in aliphatic hydrocarbons. |
| Quantity | 1 kg |
| Flash Point | >240°C (464°F) |
| Odor | Odorless |
| Chemical Name or Material | Polyethylene glycol 12,000 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.