Learn More
Kartell™ Ranvier Type Dropping Bottle
Catalog Number 12681016
Repeatedly dispense reagents accurately with a dropping bottle, an excellent alternative to a pipetting devices. Kartell™ Ranvier Type Dropping Bottle comes complete with a polyethylene tube and rubber teat for one at a time dropping or a stream.
  • Comes with PE tube and rubber teat
  • Suitable for foodstuff