Learn More
Simport™ Polyethylene Colored Screw Caps
Reduce the chances of sample contamination with these caps. Simport™ Polyethylene Colored Screw Caps are molded with a deep internal lip that fits snugly against the interior wall of the tube, and prevents the contents from coming in contact with the seal or threads.
Brand: Simport™ T340YLS
Code : 46
Additional Details : Weight : 0.29700kg
Description
- Six styles of caps to choose from and two sealing types: O-ring seal and lip seal
- High profile facilitates manipulation especially in aseptic procedures and can remain attached (T332 & T336 Series) to the tube in order to eliminate mix-ups and contamination
- All tubes and closures are manufactured in a clean environment
- Closures in natural color can be COLOR CODED by the use of T345 Series Colored Capinsert™ inserted on the top of the closure
- Certified RNase-, DNase-, Pyrogen- and DNA-free
Specifications
Screw Cap | |
Polyethylene | |
1000 Pack |
Autoclavable | |
Yellow | |
T332 & T336 Series Test Tube |