missing translation for 'onlineSavingsMsg'
Learn More

Deltalab™ Polyethylene Caps

Product Code. p-8009133
Click to view available options
Color:
Red
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12044563

Brand: Deltalab™ 307206

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Product Type Test Tube Plug Cap
Material Polyethylene
Color Red
No. per Pack 2500 Pack
Quantity 2500 Pack
For Use With (Equipment) 15 mm Tubes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.