Learn More
Polyethylene Barrel Tap
Description
Two popular sized drum taps with efficient closing for safety purposes. They are particularly appropriate where containers are stored horizontally. Constructed in high density polyethylene they are resistant to a wide range of chemicals. The materials used meet FDA requirements. The 2 inch size is particularly useful for viscous liquids and for 205L drums, whereas the 3/4 inch tap fits smaller containers. In the case of the 2 inch tap, an adapter is offered which converts the BSP thread to Tri-sure for use with containers where this thread is fitted.
Specifications
Specifications
| Description | 3/4 in. (1.905 cm) |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.