Learn More
Masterflex™ Polyethylene Barbed Y-Connectors
Product Code. 11770229
Change view
Click to view available options
Tubing Size:
1/2 in.
1/8 in.
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Size unitSize
11770229 1/8 in. Pack of 10
12848100 1/2 in. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11770229 Supplier Masterflex™ Supplier No. 3072603
Only null left
Add to basket
Add to basket
Ideal for applications such as pharmaceuticals, laboratory, industrial/manufacturing, water sampling, food processing
  • Barbed Y-Connectors
  • Polyethylene
  • Available in pack of ten
Specifications
Description Barb Connections
Autoclavable Not Autoclavable
Material HDPE
Tubing Size 1/8 in.
Hose Barb 1/8 in.
Outlet Port 1/8 in.
Port Size 1/8 in.
Sterilization Method Ethylene Oxide
Color Translucent
Product Type Barbed Y Connector