missing translation for 'onlineSavingsMsg'
Learn More

Bryson Packaging™ Polyethylene Bags

Transparent and excellent for shipping, storage and as sample bags.

Brand:  Bryson Packaging™ BAJ-300-110R

Code : 33

Additional Details : Weight : 0.27900kg

 View more versions of this product

Product Code. 12340499

  • £42.18 / Pack of 50
In Stock
to see stock.
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Description

Description

  • Available in a wide range of sizes/capacities
  • Suitable for food use
Specifications

Specifications

Bag
50 Pack
200 mm
Polyethylene
300 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers