missing translation for 'onlineSavingsMsg'
Learn More

Bryson Packaging™ Polyethylene Bags

Transparent and excellent for shipping, storage and as sample bags.

Brand:  Bryson Packaging™ BAJ-300-050J

Code : 33

Additional Details : Weight : 0.10350kg

 View more versions of this product

Product Code. 12310499

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Available in a wide range of sizes/capacities
  • Suitable for food use
Specifications

Specifications

Bag
100 Pack
150 mm
Polyethylene
100 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers