missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyester/Polypropylene Pouch
Brand: Synergy Healthcare 91BOP02550
Code : 03
Additional Details : Weight : 0.20000kg
Description
Manufactured from transparent polyester/polypropylene film for strength and puncture resistance, and high quality wet strength MG kraft paper. Opens along width. Conforms to BS 6871.Suitable for both steam and ethylene oxide sterilisation.Specifications
For strength and puncture resistance and high quality wet strength MG kraft paper | |
500Pack | |
Polyester/Polypropylene |
508mm | |
Clear | |
254mm |