missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Stainless Steel Benchtop Disposal System

Product Code. 11755084
Change view
Click to view available options
Dimensions (L x W):
190 x 190 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
11755084 190 x 190 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11755084 Supplier Bel-Art™ Supplier No. H131940011

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Keeps odors contained. Bel Art™ Polyester Benchtop Disposal System has a hinged, swinging lid that allows easy access.

  • Perfect for small items such as pipette tips and microcentrifuge tubes
  • 1.5L capacity
  • Holds a 25 x 25cm (10 x 10 in.) autoclavable biohazard disposal bag (Bel Art product #H13182-0000)
  • Aesthetic and durabl
  • Resists fingerprint smudges and stains

Specifications

Diameter (Metric) Outer 150 mm
Dimensions (L x W) 190 x 190 mm
Height (Metric) 190 mm
Length (Metric) 190 mm
Autoclavable Autoclavable
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.