missing translation for 'onlineSavingsMsg'
Learn More

Polyester acrylate/methacrylate, PEAM-645

Product Code. 15442727
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15442727

Brand: Thermo Scientific Alfa Aesar 046596.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recommended for use as a base resin in adhesive applications or coating applications | CAS: 921213-39-0

Recommended for use as a base resin in adhesive applications or coating applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 921213-39-0

Specifications

Color Amber to Brown
Physical Form Liquid
Melting Point >375°C (decomposition)
Quantity 25 g
Formula Weight 645
Odor Mild
Chemical Name or Material Polyester acrylate/methacrylate, PEAM-645

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.