missing translation for 'onlineSavingsMsg'
Learn More

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 9000, Thermo Scientific Chemicals

Catalog Number p-7052086
Click to view available options
Quantity:
50 mL
250 mL
1 L
This item is not returnable. View return policy
9016-00-6
(C2H6OSi)n
74.15
MFCD00084411
SEUDSDUUJXTXSV-UHFFFAOYSA-N
dimethylsilanone
C[Si](C)(-*)O-*
This item is not returnable. View return policy
9016-00-6
(C2H6OSi)n
74.15
MFCD00084411
SEUDSDUUJXTXSV-UHFFFAOYSA-N
dimethylsilanone
C[Si](C)(-*)O-*

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Physical Form Viscous Liquid
Boiling Point >205°C
Melting Point <-40°C
Density 0.968 g/mL
Quantity 50 mL
Flash Point 315°C (599°F)
Odor Odorless
Refractive Index 1.403
Chemical Name or Material Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 9,000

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'