Learn More
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 139,000
Description
Polydimethylsiloxane, trimethylsiloxy is used in water-repellent, protective creams & lotions. It is also used as surfactant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless |
| Physical Form | Viscous Liquid |
| Boiling Point | >205°C |
| Melting Point | <-40°C |
| Density | 0.977 g/mL |
| Solubility Information | Not miscible with water. Insoluble in methanol,and ethylene glycol. |
| Quantity | 4 kg |
| Flash Point | 321°C (610°F) |
| Odor | Odorless |
| Refractive Index | 1.4035 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.