missing translation for 'onlineSavingsMsg'
Learn More

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 1250

Product Code. p-7052082
Click to view available options
Quantity:
50 mL
250 mL
1 L
Unit Size:
1 L
250mL
50mL
This item is not returnable. View return policy

Product Code. 11349038

Brand: Thermo Scientific Alfa Aesar 042498.AD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9016-00-6 | (C2H6OSi)n | 74.15 g/mol

Polydimethylsiloxane, trimethylsiloxy terminated is widely used as a common surfactant. It is an active component of defoamers which suppress the formation of foams and as a critical ingredient in water-repelling coatings as an active silicone fluid. It finds application in optic telecommunications, biomedical research, elasticity of polymer networks, adhesive materials and in skin-moisturizing lotions. Furthermore, it is used as a filler fluid in breast implants and as a sorbent for the analysis of headspace (dissolved gas analysis) of food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9016-00-6
Molecular Formula (C2H6OSi)n
Molecular Weight (g/mol) 74.15
MDL Number MFCD00084411
InChI Key SEUDSDUUJXTXSV-UHFFFAOYSA-N
IUPAC Name dimethylsilanone
SMILES C[Si](C)(-*)O-*

Specifications

Color Colorless
Physical Form Liquid
Boiling Point >205°C
Melting Point -40°C
Density 0.935 g/mL
Solubility Information Slightly miscible with water. Immiscible with methanol and ethylene glycol.
Quantity 50 mL
Flash Point 263°C (505°F)
Odor Odorless
Refractive Index 1.399
Chemical Name or Material Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 1,250
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.