missing translation for 'onlineSavingsMsg'
Learn More

Polydimethylsiloxane, hydroxy terminated, M.W. 4200

Product Code. 11390398
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
250mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11390398 50 mL 50mL
11380398 250 mL 250mL
11370398 1 L 1 L
11360398 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11390398 Supplier Thermo Scientific Alfa Aesar Supplier No. 043769.AD
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 70131-67-8 | HOSi | 45.09 g/mol

Polydimethylsiloxane is frequently used in a liquid containing 4-5% silica gel, emulsifiers and preservatives. It is employed in applications like contact lenses, medical devices, elastomers, in shampoos, food (antifoaming agent), caulking, lubricants, kinetic sand , and heat-resistant tiles. PDMS is a common surfactant, component of defoamers and as an herbicidal penetran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70131-67-8
Molecular Formula HOSi
Molecular Weight (g/mol) 45.09
MDL Number MFCD01325012
InChI Key XQSFXFQDJCDXDT-UHFFFAOYSA-N
Synonym silanol, unii-079v3j9o3x, silanole, silanol terminated polydimethylsiloxane
PubChem CID 6327522
IUPAC Name hydroxysilicon
SMILES O[Si]

Specifications

Physical Form Liquid
Density 0.97 g/mL
Solubility Information Soluble in carbon tetrachloride,benzene,chloroform,diethyl ether,toluene and other organic solvents. Insoluble in water and ethanol.
Quantity 50 mL
Chemical Name or Material Polydimethylsiloxane, hydroxy terminated, M.W. 4,200

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.