missing translation for 'onlineSavingsMsg'
Learn More

Polydimethylsiloxane, hydroxy terminated, M.W. 4200

Product Code. 11370398
Click to view available options
Quantity:
50 mL
250 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
250mL
50mL
This item is not returnable. View return policy

Product Code. 11370398

Brand: Thermo Scientific Alfa Aesar 043769.K2

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 70131-67-8 | HOSi | 45.09 g/mol

Polydimethylsiloxane is frequently used in a liquid containing 4-5% silica gel, emulsifiers and preservatives. It is employed in applications like contact lenses, medical devices, elastomers, in shampoos, food (antifoaming agent), caulking, lubricants, kinetic sand , and heat-resistant tiles. PDMS is a common surfactant, component of defoamers and as an herbicidal penetran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70131-67-8
Molecular Formula HOSi
Molecular Weight (g/mol) 45.09
MDL Number MFCD01325012
InChI Key XQSFXFQDJCDXDT-UHFFFAOYSA-N
Synonym silanol, unii-079v3j9o3x, silanole, silanol terminated polydimethylsiloxane
PubChem CID 6327522
IUPAC Name hydroxysilicon
SMILES O[Si]

Specifications

Physical Form Liquid
Density 0.97 g/mL
Solubility Information Soluble in carbon tetrachloride,benzene,chloroform,diethyl ether,toluene and other organic solvents. Insoluble in water and ethanol.
Quantity 1 L
Chemical Name or Material Polydimethylsiloxane, hydroxy terminated, M.W. 4,200

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.