missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polydextrose, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Polydextrose |
| CAS | 68424-04-4 |
| Molecular Formula | C12H22O11 |
| Quantity | 5 g |
| Synonym | POLYDEXTROSE, Poly-D-glucose, dextrose/sorbitol condensation polymer, Polydextrose solution, Unii-vh2xou12ie, Water-soluble dietary fiber |
| InChI Formula | InChI=1S/C12H22O11/c13-1-3-5(14)8(17)10(19)12(23-3)21-2-4-6(15)7(16)9(18)11(20)22-4/h3-20H,1-2H2/t3-,4-,5-,6-,7+,8+,9-,10-,11+,12?/m1/s1 |
| SMILES | C([C@@H]1[C@H]([C@@H]([C@H](C(O1)OC[C@@H]2[C@H]([C@@H]([C@H]([C@H](O2)O)O)O)O)O)O)O)O |
| Molecular Weight (g/mol) | 342.3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction