missing translation for 'onlineSavingsMsg'
Learn More

Polydextrose

Product Code. 16607072 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16607072 25 g 25g
16697062 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16607072 Supplier Thermo Scientific Acros Supplier No. 466360250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68424-04-4 | (C6H10O5)n

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68424-04-4
Molecular Formula (C6H10O5)n
MDL Number MFCD00804499
IUPAC Name Polydextrose
SMILES OCC1OC(-*)[C@@H](O)C(O)[C@@H]1O-*

Specifications

Color White
Physical Form White to yellow-ish powder
Melting Point 110°C to 130°C
Solubility Information soluble aq. soln.
Quantity 25 g
Loss on Drying ≤4 %
Packaging glass bottle
Chemical Name or Material Polydextrose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.