missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Polycystin 2 Polyclonal antibody specifically detects Polycystin 2 in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Polycystin 2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate |
| Gene Alias | APKD2, Autosomal dominant polycystic kidney disease type II protein, MGC138466, MGC138468, Pc-2, PC2, PKD4, polycystic kidney disease 2 (autosomal dominant), Polycystic kidney disease 2 protein, polycystin-2, Polycystwin, R48321, transient receptor potential cation channel, subfamily P, member 2, TRPP2 |
| Host Species | Rabbit |
| Immunogen | maps to a region between residue 918 and 968 of human polycystin-2 using the numbering given in entry NP_000288.1 |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?