missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polycystin 2 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-30117CL1
This item is not returnable.
View return policy
Description
Polycystin 2 Polyclonal antibody specifically detects Polycystin 2 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Polycystin 2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| APKD2, Autosomal dominant polycystic kidney disease type II protein, MGC138466, MGC138468, Pc-2, PC2, PKD4, polycystic kidney disease 2 (autosomal dominant), Polycystic kidney disease 2 protein, polycystin-2, Polycystwin, R48321, transient receptor potential cation channel, subfamily P, member 2, TRPP2 | |
| maps to a region between residue 918 and 968 of human polycystin-2 using the numbering given in entry NP_000288.1 | |
| 0.1 mL | |
| Cancer, Protein Kinase, Signal Transduction, Tumor Suppressors | |
| 5311 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction