missing translation for 'onlineSavingsMsg'
Learn More

Polycizer 632, TRC

Product Code. 30180070
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180070 25 g 25g
30180051 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30180070 Supplier Toronto Research Chemicals Supplier No. TRCP69897025G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-97-5
Molecular Formula C26 H50 O4
Molecular Weight (g/mol) 426.67
Synonym Adipic acid, didecyl ester (6CI,7CI,8CI), Hexanedioic acid, didecyl ester (9CI), DB 31, DB 31 (ester), Di-n-Decyl adipate, Dicapryl adipate, Didecyl adipate, NSC 4445, Polycizer 632
IUPAC Name didecyl hexanedioate
SMILES CCCCCCCCCCOC(=O)CCCCC(=O)OCCCCCCCCCC

Specifications

Quantity 25 g
InChI Formula InChI=1S/C26H50O4/c1-3-5-7-9-11-13-15-19-23-29-25(27)21-17-18-22-26(28)30-24-20-16-14-12-10-8-6-4-2/h3-24H2,1-2H3
Formula Weight 426.3709
Chemical Name or Material Adipic acid, didecyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.