missing translation for 'onlineSavingsMsg'
Learn More

Polycarbonate resin, approx. M.W. 45.000, pellets

Product Code. 10163810 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
250 g
500 g
Packaging:
Glass bottle
Unit Size:
250g
500g
5g
This item is not returnable. View return policy

Product Code. 10163810

Brand: Thermo Scientific Acros 178312500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24936-68-3 | (C16H14O3)n | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24936-68-3
Molecular Formula (C16H14O3)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00084476
SMILES CC(C)(C1=CC=C(-*)C=C1)C1=CC=C(OC(=O)O-*)C=C1

Specifications

Color Undesignated
Physical Form Pellets
Packaging Glass bottle
Density 1.2000g/mL
Quantity 250 g
Linear Formula [-C6H4C(CH3)2C6H4OCO2-]n
Chemical Name or Material Polycarbonate resin, M.W. 64.000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.