missing translation for 'onlineSavingsMsg'
Learn More

Polycarbonate resin, approx. M.W. 45.000, pellets

Product Code. 10255660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
500 g
Packaging:
Glass bottle
Unit Size:
250g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10255660 500 g Glass bottle 500g
10307840 5 g Glass bottle 5g
10163810 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10255660 Supplier Thermo Scientific Acros Supplier No. 178315000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24936-68-3 | (C16H14O3)n | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24936-68-3
Molecular Formula (C16H14O3)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00084476
SMILES CC(C)(C1=CC=C(-*)C=C1)C1=CC=C(OC(=O)O-*)C=C1

Specifications

Color Undesignated
Physical Form Pellets
Packaging Glass bottle
Density 1.2000g/mL
Quantity 500 g
Linear Formula [-C6H4C(CH3)2C6H4OCO2-]n
Chemical Name or Material Polycarbonate resin, M.W. 64.000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.