missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Polycarbonate/Polypropylene Pipette Tip
Description
- Made of Polycarbonate/Polypropylene
- Tip for micropipette
- Supplied in rack
- Compatability varies
Specifications
Specifications
| Description | Pipette tips |
| Volume (Metric) | 100 to 1000 μL |
| Color | Blue |
| Certifications/Compliance | CE/IVD |
| Tip Style | Universal |
| Packaging | Rack |
| Product Type | Pipette Tip |
| For Use With (Equipment) | Kartell, Gilson, Eppendorf, Socorex, Finnpipette, Brand, Hamilton |
| Material | Polycarbonate/Polypropylene |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?