Learn More
Nickel Electro™ Polycarbonate Hinged Gable Lid
Product Code. 11786577 Shop All Nickel-Electro Products
Change view
Click to view available options
For Use With (Equipment):
22 and 28 L Unstirred/Shaking Baths
8 and 10 L Unstirred/Shaking Baths
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
11786577 22 and 28 L Unstirred/Shaking Baths Each
12758888 8 and 10 L Unstirred/Shaking Baths Each
2 options
Product Code. 11786577
Brand: Nickel Electro™ GL122
Only null left
Add to basket
Add to basket
Maximize bath area with a hinged lid that drips condensation back into bath. Nickel Electro™ Polycarbonate Hinged Gable Lid are transparent to allows samples to be viewed.
  • Lightweight
  • Semi-transparent for viewing samples
  • Maximize bath area
Specifications
For Use With (Equipment) 22 and 28 L Unstirred/Shaking Baths
For Use With (Application) Reducing Water Evaporation, Keeping Samples and Water Free From Contamination and Saves Energy
Material Polycarbonate