missing translation for 'onlineSavingsMsg'
Learn More

Polycarbamate, TRC

Product Code. 30424248
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424248 10 mg 10mg
30417744 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30424248 Supplier Toronto Research Chemicals Supplier No. TRCP69896010MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64440-88-6
Molecular Formula C10 H18 N4 S8 Zn2
Molecular Weight (g/mol) 581.61
Synonym Zinc, bis(N,N-dimethylcarbamodithioato-κS,κS')[μ-[[N,N'-1,2-ethanediylbis[carbamodithioato-κS,κS']](2-)]]di-, Zinc, bis(dimethylcarbamodithioato-S,S')[μ-[[1,2-ethanediylbis[carbamodithioato]](2-)]]di-, Zinc, bis(dimethylcarbamodithioato-κS,κS')[μ-[[1,2-ethanediylbis[carbamodithioato-κS,κS']](2-)]]di- (9CI), Bis(N,N-dimethylcarbamodithioato-κS,κS')[μ-[[N,N'-1,2-ethanediylbis[carbamodithioato-κS,κS']](2-)]]dizinc, Carbamodithioic acid, 1,2-ethanediylbis-, zinc complex, Carbamodithioic acid, dimethyl-, zinc complex, Bis-Dithane, Bisdimethyldithiocarbamoyl zinc ethylenebis(dithiocarbamate), Polycarbamate, TOC 3204, TOC 3204F
IUPAC Name dimethylcarbamothioylsulfanyl-[2-(dimethylcarbamothioylsulfanylzinciosulfanylcarbothioylamino)ethylcarbamothioylsulfanyl]zinc
SMILES CN(C)C(=S)S[Zn]SC(=S)NCCNC(=S)S[Zn]SC(=S)N(C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C4H8N2S4.2C3H7NS2.2Zn/c7-3(8)5-1-2-6-4(9)10;2*1-4(2)3(5)6;;/h1-2H2,(H2,5,7,8)(H2,6,9,10);2*1-2H3,(H,5,6);;/q;;;2*+2/p-4
Formula Weight 577.788
Chemical Name or Material Polycarbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.