missing translation for 'onlineSavingsMsg'
Learn More

Polycaprolactone, M.W. approx. 80,000

Product Code. 17231152 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17231152 500 g 500g
17138258 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17231152 Supplier Thermo Scientific Acros Supplier No. 178305000
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24980-41-4 | (C6H10O2)n | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24980-41-4
Molecular Formula (C6H10O2)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00084404
InChI Key XNCNNDVCAUWAIT-UHFFFAOYSA-N
IUPAC Name oxepan-2-one
SMILES *-CCCCCC(=O)O-*

Specifications

Melting Point 60.0°C
Linear Formula [-O(CH2)5CO-]n
Quantity 500 g
Chemical Name or Material Polycaprolactone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.