missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ PolyBlock for 3 x 28 mm Reagent Vials

Product Code. 11302773 Shop All Heidolph Instruments Products
Change view
Click to view available options
For Use With (Equipment):
StarFish Multi-Experiment Workstation
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11302773 StarFish Multi-Experiment Workstation Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11302773 Supplier Heidolph™ Supplier No. 5058002500
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

PolyBlock holding three 28 mm reagent vials for use with the StarFish Multi-Experiment Workstation.

Along with the StarFish base plate, the PolyBlocks are among the basic elements of the entire StarFish Workstation. The PolyBlocks are attachment segments that can be combined for reaction vessels of different sizes (up to 5 different inserts per StarFish system).

  • Chemical-resistant attachment segment holds up to 3 reagent vessels with a diameter of 28 mm
  • Segment is suitable for heating temperatures up to 260°C
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) StarFish Multi-Experiment Workstation
Holds 3 x 28 mm Reagent Vials
Material Aluminum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.