missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Polyacetal Hose Clips

Product Code. 10382482 Shop All Bürkle Products
Change view
Click to view available options
Tubing Size:
10 to 11 mm
11 to 12 mm
12 to 14 mm
13 to 15 mm
15 to 17 mm
17 to 19 mm
18 to 20 mm
20 to 22 mm
23 to 25 mm
6 to 6.5 mm
9 to 10 mm
Unit Size:
Pack of 10
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Size Unit Size
10382482 6 to 6.5 mm Pack of 10
10054083 9 to 10 mm Pack of 10
10591954 10 to 11 mm Pack of 10
10696453 11 to 12 mm Pack of 10
10542403 12 to 14 mm Pack of 10
10347752 13 to 15 mm Pack of 10
10275872 15 to 17 mm Pack of 10
10163042 17 to 19 mm Pack of 10
10616843 18 to 20 mm Pack of 10
10001922 20 to 22 mm Pack of 10
10033513 23 to 25 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Product Code. 10382482 Supplier Buerkle™ Supplier No. 86780006
Sample Preparation and Storage Essentials
Use promo code 27996
View all promotions
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use these Buerkle™ Polyacetal Hose Clips to clamp down on tubing for a sure and secure fit.

  • Hose clips made of Polyacetal (POM)
  • Featured in varying diameters
  • POM is white, sterilizable, does not rust, permanently elastic and hygienic.
TRUSTED_SUSTAINABILITY

Specifications

Material Polyacetal
For Use With Hoses
Packaging Quantity 10 Pc./Pk.
Tubing Size 6 to 6.5 mm
Color White
Product Type Hose Clip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.