missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinylbenzyl chloride)

Product Code. 11364718
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364718 50 g 50g
11336189 2 g 2g
11374718 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11364718 Supplier Thermo Scientific Alfa Aesar Supplier No. 043067.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9080-67-5 | −[CH2CH(C6H4CH2Cl-]n

In scientific and educational research. In synthesis of polymer supported reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9080-67-5
Molecular Formula −[CH2CH(C6H4CH2Cl-]n
MDL Number MFCD00135445

Specifications

Physical Form Solid
Solubility Information Insoluble in water.
Quantity 50 g
Chemical Name or Material Poly (vinylbenzyl chloride)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.