missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Poly(vinyl formal)

Brand:  Alfa Aesar™ 044190.14

Code : NEW

Additional Details : CAS Number : 9003-33-2 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11339688

  • £13.90 / 25g


Estimated Shipment date
from Supplier 21-02-2018
Add to basket

Specifications

Chemical Identifiers

Poly (vinyl formal)
9003-33-2
MFCD00084463

Specifications

Poly (vinyl formal)
1.23g/mL
MFCD00084463
25g
9003-33-2
Granular Powder
Soluble in acetone, chloroform, dioxane, THF
Videos
SDS