missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl formal)

Product Code. 11349688
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349688 500 g 500g
11339688 25 g 25g
11359688 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11349688 Supplier Thermo Scientific Alfa Aesar Supplier No. 044190.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 9003-33-2 | (C5H8O2)n | 100.12 g/mol

It is used in the anomalous sorption and pervaporation of aqueous organic mixtures by poly(vinyl acetal) membranes. Poly(vinyl formal) is mainly used in three general applications wire coatings, surface coatings and adhesives. The first and extensive application of poly(vinyl formal) is in the manufacture of a tough, adhesive and heat resistant wire enamel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-33-2
Molecular Formula (C5H8O2)n
Molecular Weight (g/mol) 100.12
MDL Number MFCD00084463
InChI Key DFFZYNIDEOOVAU-UHFFFAOYSA-N
Synonym poly vinyl formal, ethenyloxy methoxy ethene, polyvinyl formal, ethene, 1,1'-methylenebis oxy bis-, homopolymer, vinyloxymethoxyethylene, methylene glycol divinyl ether, 1,1'-methylenebisoxy bisethene, ethene,1,1'-methylenebis oxy bis-9ci
PubChem CID 165717
IUPAC Name ethenoxymethoxyethene
SMILES *-CC1CC(-*)OCO1

Specifications

Physical Form Granular Powder
Density 1.23g/mL
Solubility Information Soluble in acetone, chloroform, dioxane, THF
Quantity 500 g
Chemical Name or Material Poly (vinyl formal)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.