missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Poly(vinyl formal)

Brand:  Alfa Aesar™ 044190.22

Code : NEW

Additional Details : CAS Number : 9003-33-2 Weight : 0.10000kg

Quantity 100g

 View more versions of this product

Product Code. 11359688

  • £39.90 / 100g


EU Stock 2
Add to basket

Specifications

Chemical Identifiers

Poly (vinyl formal)
9003-33-2
MFCD00084463

Specifications

Poly (vinyl formal)
1.23g/mL
MFCD00084463
100g
9003-33-2
Granular Powder
Soluble in acetone, chloroform, dioxane, THF
Videos
SDS