missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl alcohol), Honeywell™ Fluka™

Product Code. 15627170 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15627170 100 g Poly bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15627170 Supplier Honeywell-Fluka Supplier No. 81366100G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Protective colloid for titrations

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 9002-89-5
Molecular Formula (C2H4O)n
Linear Formula [-CH2CHOH-]n
MDL Number MFCD00081922
UN Number NONH for all modes of transport
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
Quantity 100 g
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
SMILES OC(-*)C-*
IUPAC Name ethenol
Molecular Weight (g/mol) 44.05
PubChem CID 11199
Packaging Poly bottle
Chemical Name or Material Poly(vinyl alcohol)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.