missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl alcohol), 98% hydrolyzed, average M.W. 16000, ACROS Organics™

Product Code. 10032420 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10032420

Brand: Thermo Scientific Acros 183300250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
PubChem CID 11199
IUPAC Name ethenol
SMILES OC(-*)C-*

Specifications

Color White
Physical Form Powder or Granules
Ash 1% max.
Linear Formula [-CH2CH(OH)-]n
Merck Index 15, 7706
Quantity 25 g
Percent Purity 98% min. (degree of hydrolysis)
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Glass bottle
Viscosity 6mPa·s (20°C 4% aq. solution)
Chemical Name or Material Poly(vinyl alcohol)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.