missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl alcohol), 87.0-89.0% hydrolyzed, M.W. approx. 13,000-23,000, Thermo Scientific Chemicals

Product Code. 15274208
Click to view available options
1kg; Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow to Brown
pH 4.5 to 6.5 (4% aq.sol)
CAS 9002-89-5
Physical Form Liquid After Melting
Melting Point 230.0°C to 240.0°C
Ash 1% max. (as Na2O)
Molecular Formula (C2H4O)n
Linear Formula [-CH2CH(OH)-]n
MDL Number MFCD00081922
Synonym vinyl alcohol, polyvinyl alcohol, hydroxyethene, hydroxyethylene, ethenol, homopolymer, gohsenol, polyviol, elvanol, mowiol, poval
Solubility Information Solubility in water: soluble in hot water.
Quantity 1 kg
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
SMILES OC(-*)C-*
IUPAC Name ethenol
Molecular Weight (g/mol) 44.05
PubChem CID 11199
Percent Purity 87.0 to 89.0 mol% (degree of hydrolysis)
Infrared Spectrum Authentic
Loss on Drying 5% max.
Packaging Plastic bottle
Viscosity 3.0-3.7 mPa.s (20°C 4% aq.sol.)
Chemical Name or Material Poly(vinyl alcohol), 88% Hydrolyzed, Average M.W. 22000
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'