missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl acetate), approx. M.W. 170,000

Product Code. 10604882 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
500 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
25g
500g
This item is not returnable. View return policy

Product Code. 10604882

Brand: Thermo Scientific Acros 183250010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9003-20-7 | (C4H6O2)n | 86.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-20-7
Molecular Formula (C4H6O2)n
Molecular Weight (g/mol) 86.09
MDL Number MFCD00084457
InChI Key XTXRWKRVRITETP-UHFFFAOYSA-N
Synonym vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
PubChem CID 7904
ChEBI CHEBI:46916
IUPAC Name ethenyl acetate
SMILES CC(=O)OC(-*)C-*

Specifications

Physical Form Beads
Linear Formula [-CH2CH(O2CCH3)-]n
Density 1.1800g/mL
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in low molecular weight alcohols, esters,, benzene and chorinated hydrocarbons
Quantity 1 kg
Infrared Spectrum Authentic
Loss on Drying 4% max.
Packaging Glass bottle
Specific Gravity 1.18
Chemical Name or Material Poly(vinyl acetate), Average M.W. 170.000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.