missing translation for 'onlineSavingsMsg'
Learn More

Poly(vinyl acetate), ∽ M.W. 170,000, Thermo Scientific Chemicals

Catalog Number p-186533
16.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
500 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
9003-20-7
(C4H6O2)n
86.09
MFCD00084457
XTXRWKRVRITETP-UHFFFAOYSA-N
vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
7904
CHEBI:46916
ethenyl acetate
CC(=O)OC(-*)C-*
This item is not returnable. View return policy
9003-20-7
(C4H6O2)n
86.09
MFCD00084457
XTXRWKRVRITETP-UHFFFAOYSA-N
vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
7904
CHEBI:46916
ethenyl acetate
CC(=O)OC(-*)C-*

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Beads
Linear Formula [-CH2CH(O2CCH3)-]n
Density 1.1800g/mL
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in low molecular weight alcohols, esters,, benzene and chorinated hydrocarbons
Quantity 25 g
Infrared Spectrum Authentic
Loss on Drying 4% max.
Packaging Glass bottle
Specific Gravity 1.18
Chemical Name or Material Poly(vinyl acetate), Average M.W. 170.000

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'