missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poly-Ubiquitin Antibody - K29 Linkage - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05664
This item is not returnable.
View return policy
Description
Poly-Ubiquitin Polyclonal antibody specifically detects Poly-Ubiquitin in All samples. It is validated for Western Blot, Dot Blot
Specifications
| Poly-Ubiquitin | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Dot Blot 1:500 - 1:1000 | |
| CEP80, HEL112, ribosomal protein S27a, RPS27A, S27A, UBA80, UBC, UBCEP1, UBCEP80 | |
| A synthetic peptide corresponding to a sequence containing polyubiquitinated protein (K29 linkage). | |
| 100 μg | |
| Primary | |
| All species | |
| Purified |
| Western Blot, Dot Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 6233 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction