missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Poly-Ubiquitin Antibody (Genentech patent anti-Polyubiquitin), Novus Biologicals™
Shop All Bio Techne ProductsDescription
Poly-Ubiquitin Monoclonal antibody specifically detects Poly-Ubiquitin in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
Specifications
| Antigen | Poly-Ubiquitin |
| Applications | Flow Cytometry, ELISA, Functional Assay |
| Classification | Monoclonal |
| Clone | Genentech patent anti-Polyubiquitin |
| Conjugate | Unconjugated |
| Formulation | Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) |
| Gene Accession No. | P0CG48 |
| Gene Alias | CEP80, HEL112, ribosomal protein S27a, RPS27A, S27A, UBA80, UBC, UBCEP1, UBCEP80 |
| Host Species | Human |
| Immunogen | Polyubiquitin |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?