missing translation for 'onlineSavingsMsg'
Learn More

Poly(tetrafluoroethylene)

Product Code. 10173810 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10173810 5 g Glass Bottle 5g
10042400 100 g Plastic bottle 100g
10183810 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10173810 Supplier Thermo Scientific Acros Supplier No. 178930050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9002-84-0 | (C2F4)n | NaN g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9002-84-0
Molecular Formula (C2F4)n
Molecular Weight (g/mol) NaN
MDL Number MFCD00084455
Synonym PTFE
SMILES FC(F)(-*)C(F)(F)-*

Specifications

Color White
Physical Form Powder
Linear Formula (-CF2CF2-)n
Density 2.0000g/mL
Quantity 5 g
Infrared Spectrum Authentic
Loss on Drying 1% max.
Packaging Glass Bottle
Chemical Name or Material Poly(tetrafluoroethylene)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.