missing translation for 'onlineSavingsMsg'
Learn More

Poly(styrene sulfonic acid) sodium salt, M.W. 70,000

Product Code. 15466375
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15466375 250 g 250g
15456375 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15466375 Supplier Thermo Scientific Alfa Aesar Supplier No. 041688.30
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25704-18-1 | (C8H7NaO3S)n | 184.21 g/mol

Poly(styrene sulfonic acid) sodium salt is used as Polyelectrolyte. It is also used as electroconductive and antistatic resin for electrographic and electrophotographic substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25704-18-1
Molecular Formula (C8H7NaO3S)n
Molecular Weight (g/mol) 184.21
MDL Number MFCD00084449
InChI Key MAGFQRLKWCCTQJ-UHFFFAOYSA-N
Synonym 4-styrenesulfonic acid sodium salt
PubChem CID 131675404
IUPAC Name 4-ethenylbenzenesulfonic acid;molecular hydrogen;sodium
SMILES [Na]OS(=O)(=O)C1=CC=C(C=C1)C(-*)C-*

Specifications

Quantity 250 g
Solubility Information Soluble in water,lower alcohols,glycols.
Physical Form Powder
Chemical Name or Material Poly(styrene sulfonic acid) sodium salt, M.W. 70,000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.