missing translation for 'onlineSavingsMsg'
Learn More

Poly(styrene sulfonic acid), M.W. 75,000, 30% w/v aq. soln.

Product Code. 11345377
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11345377 500 g 500g
11335377 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11345377 Supplier Thermo Scientific Alfa Aesar Supplier No. 042653.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 28210-41-5

Poly(styrene sulfonic acid) is used in water softening and to remove hard ions like calcium and magnesium. In the medical field, it is used as potassium binders in acute and chronic kidney disease. Its sodium salt is used as a superplastifier in cement, as a dye improving agent for cotton, as proton exchange membranes in fuel cell applications. It acts as a solid acid catalyst in organic synthesis. It is an antistatic resin used for electrographic and electrophotographic substrates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28210-41-5
MDL Number MFCD00165973
Synonym 4-styrenesulfonic acid, polystyrene sulfonate, tolevamer, styrene-4-sulphonic acid, 4-vinylbenzenesulfonic acid, benzenesulfonic acid, 4-ethenyl, poly 4-vinylbenzenesulfonic acid, tolevamer inn, poly styrene sulfonic acid sodium salt, unii-zsl2fb6gxn

Specifications

Color Amber
Density 1.1 g/mL
Quantity 500 g
UN Number UN2584
Solubility Information Miscible with water,ethanol,methanol,lower alcohols and glycols.
Concentration or Composition (by Analyte or Components) 30% w/v aq. soln.
Physical Form Liquid
Chemical Name or Material Poly (styrene sulfonic acid), M.W. 75,000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.