missing translation for 'onlineSavingsMsg'
Learn More

Poly(styrene-divinylbenzene), aminomethylated, 1% cross-linked, 200-400 mesh, Thermo Scientific™

Catalog Number 11428590
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
89551-24-6
C7H9N
107.156
MFCD00146442
WGQKYBSKWIADBV-UHFFFAOYSA-N
benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine
7504
CHEBI:40538
phenylmethanamine
C1=CC=C(C=C1)CN
This item is not returnable. View return policy
89551-24-6
C7H9N
107.156
MFCD00146442
WGQKYBSKWIADBV-UHFFFAOYSA-N
benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine
7504
CHEBI:40538
phenylmethanamine
C1=CC=C(C=C1)CN

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Physical Form Powder
Quantity 5 g
Mesh Size 200 to 400 Mesh
Chemical Name or Material Poly (styrene-divinylbenzene), Aminomethylated, 1% cross-linked
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'