missing translation for 'onlineSavingsMsg'
Learn More

Poly(styrene-divinylbenzene), aminomethylated, 1% cross-linked, 100-200 mesh

Product Code. 11478580
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478580 5 g 5g
11458580 25 g 25g
11468580 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478580 Supplier Thermo Scientific Alfa Aesar Supplier No. 044392.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Formation of amide groups using Boc strategy, as a scavenger resin | CAS: 89551-24-6 | C7H9N | 107.156 g/mol

Poly(styrene-divinylbenzene), aminomethylated, 1% cross-linked, 100-200 mesh is used in the formation of amide groups using Boc strategy. as a scavenger resin. C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89551-24-6
Molecular Formula C7H9N
Molecular Weight (g/mol) 107.156
MDL Number MFCD00146442
InChI Key WGQKYBSKWIADBV-UHFFFAOYSA-N
Synonym benzylamine, benzenemethanamine, monobenzylamine, alpha-aminotoluene, phenylmethyl amine, aminomethyl benzene, 1-phenylmethanamine, moringine, n-benzylamine, phenylmethylamine
PubChem CID 7504
ChEBI CHEBI:40538
IUPAC Name phenylmethanamine
SMILES C1=CC=C(C=C1)CN

Specifications

Physical Form Powder
Solubility Information Slightly soluble in water.
Quantity 5 g
Mesh Size 100 to 200 Mesh
Chemical Name or Material Poly (styrene-divinylbenzene), Aminomethylated, 1% cross-linked

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.