missing translation for 'onlineSavingsMsg'
Learn More

Poly(propylene glycol), average M.W. 2.000

Product Code. 10256963 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
500g
5g
Product Code. Quantity Packaging unitSize
10256963 2.5 kg Glass bottle 2.5kg
10489890 5 g Glass bottle 5g
10740701 500 g Glass bottle 500g
3 options
This item is not returnable. View return policy

Product Code. 10256963

Brand: Thermo Scientific Acros 192180025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25322-69-4 | (C3H6O)n | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25322-69-4
Molecular Formula (C3H6O)n
Molecular Weight (g/mol) 134.18
MDL Number MFCD00084446
InChI Key DNIAPMSPPWPWGF-UHFFFAOYNA-N
Synonym polypropylene glycol, polyoxypropylene, 2-2-hydroxypropoxy propan-1-ol, emkapyl, lineartop e, niax ppg, polyoxypropylene glycol, methyloxirane homopolymer, desmophen 360c, 2-2-hydroxypropoxy-1-propanol
PubChem CID 32881
IUPAC Name 2-(2-hydroxypropoxy)propan-1-ol
SMILES [H]OC(C)CO

Specifications

Color White to Gray
Physical Form Crystals or Crystalline Powder
Acid Value 0.1 mg KOH/g max.
Boiling Point 200.0°C
Melting Point -31.0°C
Linear Formula H[OCH(CH3)CH2]nOH
Density 1.0000g/mL
Solubility Information Solubility in water: practically insoluble.
Quantity 2.5 kg
Flash Point 230°C
Hydroxyl Value 53 to 58 mg KOH/g
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4495 to 1.4515
Specific Gravity 1
Viscosity 300 mm2/s (25°C)
Water 0.15% Max.
Chemical Name or Material Poly(propylene glycol), Average M.W. 2.000
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.