missing translation for 'onlineSavingsMsg'
Learn More

Poly(phenylene sulfide), powder, nominal M.W. 10000, ACROS Organics™

Brand:  Acros Organics 178852500

Code : 29

Additional Details : CAS Number : 9016-75-5 Weight : 0.30000kg

Quantity 250g
Packaging Plastic bottle

 View more versions of this product

Product Code. 10002370

  • £110.70 / 250g


Estimated Shipment
19-03-2018
Add to basket

Specifications

Chemical Identifiers

Poly(phenylene sulfide)
9016-75-5
MFCD00084445
nominal M.W. 10000
(-C6H4S-)n

Specifications

Poly(phenylene sulfide)
9016-75-5
Powder
MFCD00084445
1% max.
1.36
250g
nominal M.W. 10000
1.3600g/mL
(-C6H4S-)n
Authentic
Plastic bottle
Off-white to tan
Videos
SDS