missing translation for 'onlineSavingsMsg'
Learn More

Poly(methylhydrosiloxane)

Product Code. 11363586
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363586 50 g 50g
11373586 250 g 250g
11383586 1000 g 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11363586 Supplier Thermo Scientific Alfa Aesar Supplier No. L14561.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 63148-57-2 | (CH4OSi)n

Poly(methylhydrosiloxane) is used in the reduction of esters to alcohols, catalyzed by a combination of titanocene dichloride and either n-BuLi or EtMgBr. It is a safer alternative to triethoxysilane, B22063, for reduction of phosphine oxides to phosphines, catalyzed by Ti(O-i-Pr)4. It is also used in greener amine synthesis by reductive amination as an alternative to borohydrides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63148-57-2
Molecular Formula (CH4OSi)n
MDL Number MFCD00084478
InChI Key QNWOFLWXQGHSRH-UHFFFAOYSA-N
Synonym Methyl hydrogen polysiloxane; PMHS
SMILES C[SiH](O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Density 1.006
Solubility Information Not miscible or difficult to mix in water.
Quantity 50 g
Formula Weight ca 1900
Flash Point 204°C (399°F)
Refractive Index 1.398
Chemical Name or Material Poly(methylhydrosiloxane)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.