missing translation for 'onlineSavingsMsg'
Learn More

Poly(methyl Methacrylate), TRC

Product Code. 30012630
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012630 5 g 5g
30135206 2.5 g 2.5g
30004859 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012630 Supplier Toronto Research Chemicals Supplier No. TRCP6880985G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9011-14-7
Formula Weight 300.1573
Molecular Formula C15 H24 O6 X2
Molecular Weight (g/mol) 300.35
SMILES [X]CC(C)(CC(C)(CC([X])(C)C(=O)OC)C(=O)OC)C(=O)OC
Synonym PMMA
Chemical Name or Material Polymethyl methacrylate

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.